Mouse Recombinant IFN-G (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
200-16-100UG 200-16-1MG 200-16-20UG 200-16-500UG 200-16-250UG
10787-950EA 360.86 CAD
10787-950 10787-952 10787-954 10821-828 10822-054
Mouse Recombinant IFN-G (from E. coli)
Proteins and Peptides
Interferon gamma (IFN-γ) is a type II interferon that is critical during adaptive and innate immune responses to infection. IFN-γ is produced by T cells and natural killer cells following antigen-specific activation. IFN-γ binds IFN-γ receptors (IFN-γ R1 and IFN-γ R2), which are expressed on most immune cells, to activate the JAK-STAT pathway. IFN-γ-induced signaling increases the expression of class 1 major histocompatibility complex (MHC) molecules. Mouse IFN-γ is not cross-reactive with human IFN-γ.
  • High quality
  • Low endotoxin
  • Affordable
Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.
Caution: For research use only.