ART®, Filter Tips, Thermo Scientific
Supplier: Thermo Scientific
2330-05 2350
72830-556CS 2154.66 CAD
72830-556 72830-562
ART®, Filter Tips, Thermo Scientific
Pipette Tips
Manufactured using a special process, tips are furnished with a unique ART® filter to eliminate any risk of cross-contamination between samples, which is particularly important in molecular biology. They are suitable for applications involving the use of DNA or RNA, as well as radioactive and infectious samples.
  • Certified free of RNase, DNase, DNA and pyrogens
  • Sterile tips: Sterilised by gamma radiation
  • 'Low Retention' tips: Extremely hydrophobic surface for lower retention, superior precision and reproducibility of measurements
Ordering information:More Product Information