Mouse Recombinant SCF (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
200-09-1MG 200-09-100UG 200-09-10UG 200-09-500UG
10787-900EA 6524.5 CAD
10787-900 10787-902 10788-760 10788-762 10822-364
Mouse Recombinant SCF (from E. coli)
Proteins and Peptides
Stem cell factor (SCF) is a cytokine made by fibroblasts and endothelial cells. SCF binds to the receptor c-Kit/CD117 and plays a critical role in the maintenance, survival, and differentiation of hematopoietic stem cells. While human SCF shows no activity on murine cells, murine and rat SCF are active on human cells.
  • High quality
  • Low endotoxin
  • Affordable
Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.
Caution: For research use only.