Acids and Bases Kit
Supplier: ALDON CORP SE
IS8006
470163-078KT 55.6 CAD
470163-078
Acids and Bases Kit
Educational Materials Chemistry Educational Materials Acids and Bases Learning Activities
Use Standardization Techniques to Determine Molarity
  • Classic AP Chemistry Experiment
  • Instructions Included
A solution of sodium hydroxide which is approximately 0.01M will be standardized with a solid acid, potassium hydrogen phthalate, to determine the exact Molarity of the base. The standardized base will then be titrated against a monoprotic unknown acid to determine the molarity of the acid. A pH indicator and/or a pH meter will be used to determine the equivalence point.
Ordering information: Includes Hydrochloric Acid, Sodium Hydroxide, Potassium Hydrogen Phthalate, and Phenolphthalein.