Mouse Recombinant M-CSF (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
200-08-100UG 200-08-10UG 200-08-1MG 200-08-500UG
10787-888EA 1145.07 CAD
10787-888 10787-890 10787-892 10787-894 10822-268
Mouse Recombinant M-CSF (from E. coli)
Proteins and Peptides
Macrophage colony stimulating factor (M-CSF) is a hematopoietic growth factor that is widely produced by a variety of cells. M-CSF stimulates the proliferation and differentiation of hematopoietic stem cells into monocyte and macrophage cell types. M-CSF also acts through the colony stimulating factor 1 receptor (CSF1R) to modulate processes involved in immunology, bone metabolism, fertility, and pregnancy. Human M-CSF shows activity on mouse cells; however, mouse M-CSF shows no activity on human cells.
  • High quality
  • Low endotoxin
  • Affordable
Caution: For research use only.