GHS Label Rolls "Mini Pictograms"
Supplier: EMEDCO
GHMLRICRQ34 GHMLRITOQ34 GHMLRICBQ34 GHMLRIENQ34 GHMLRIEXQ34 GHMLRIGZQ34 GHMLRIINQ34 GHMLRIDAQ34 GHMLRIMUQ34
470165-248RL 382.85 CAD
470165-248 470165-250 470165-252 470165-256 470165-258 470165-260 470165-314 470165-316 470165-318
GHS Label Rolls "Mini Pictograms"
Labels
GHS Pictogram labels help identify hazardous items throughout the facility.
Pre-printed, die-cut GHS labels stick to any kind of surface. GHS pictogram labels do not peel or tear. They have a white background with a red border and black graphics.
The labels comply with OSHA standards.
Packaging: One roll has 500 labels.