Anti-the Tyrosine Kinase Receptor B ECD Rabbit Polyclonal Antibody

Supplier: Biosensis
R-149-100
10782-482EA 542.49 CAD
10782-482
Anti-the Tyrosine Kinase Receptor B ECD Rabbit Polyclonal Antibody
Antibodies
TrkB is a member of the neurotrophic tyrosine receptor kinase family. It is a membrane-bound receptor and upon neurotrophin binding, it phosphorylates itself as well as MAPK pathways members. TrkB is the receptor for brain-derived neurotrophic factor (BDNF), neurotrophin-3 and neurotrophin-4/5 but not nerve growth factor (NGF). It is Involved in the development and/or maintenance of the nervous system. SUBUNIT: Exists in a dynamic equilibrium between monomeric (low affinity) and dimeric (high affinity) structures.  SUBCELLULAR LOCATION: Membrane; single-pass type I membrane protein. ALTERNATIVE PRODUCTS: 4 named isoforms produced by alternative splicing. Additional isoforms seem to exist. TISSUE SPECIFICITY: The different forms are differentially expressed in various cell types. SIMILARITY: Belongs to the Tyr protein kinase family. Insulin receptor subfamily. SIMILARITY: Contains 2 Ig-like C2-type (immunoglobulin-like) domains. SIMILARITY: Contains 2 LRR (leucine-rich) repeats. SIMILARITY: Contains 1 protein kinase domain. Mutations in the TrkB gene have been associated with obesity and mood disorders.

Specificity was demonstrated by immunohistochemistry. This antibody was used to stain cryostat sections of the rat peripheral sensory ganglia.
Application Information:
IHC, 1-site ELISA (1:10000 dilution). A dilution of 1:1000 to 1:3000 is recommended for IHC. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: the Tyrosine Kinase Receptor B ECD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Multiple points of reactivity likely because of the polyclonal nature of the antibody
Host: Rabbit
Isotype: IgG
Reactivity: Rat, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR