Anti-Myc Tag Rabbit Polyclonal Antibody

Supplier: Biosensis
R-1319-100
10782-450EA 674.66 CAD
10782-450
Anti-Myc Tag Rabbit Polyclonal Antibody
Antibodies
The Myc tag contains the amino acids Glu-Gln-Lys-Leu-Ile-Ser-Glu-Glu-Asp-Leu (E-Q-K-L-I-S-E-E-D-L) corresponding to amino acids 410-419 of human Myc. This tag is widely used for monitoring expression of recombinant proteins in bacteria, insect and mammalian cells.

This polyclonal anti-Myc-tag antibody detects overexpressed proteins containing the Myc epitope tag. The antibody recognizes the Myc-tag fused to either the amino- or carboxy- termini of targeted proteins in transfected or transformed cells.
Application Information:
ELISA and Western Blotting (WB). Suggested dilutions for ELISA at 1:1,000 and WB 1:500 - 1:1,000. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Tag
Antigen: Myc Tag
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Myc epitope
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR