Anti-PS2 Chicken Polyclonal Antibody

Supplier: Biosensis
C-1535-500
10781-968EA 516.66 CAD
10781-968
Anti-PS2 Chicken Polyclonal Antibody
Antibodies
Presenilin-2 (PSEN2) is a multi-pass membrane protein and component of the gamma-secretase complex. Defects in PSEN2 are a cause of Alzheimer disease type 4 (AD4), an autosomal dominant Alzheimer disease. (Ref:SWISS-Prot).

Antibody recognizes Presenillin-2 in western blot and by ELISA, other applications have not yet been tested.
Application Information:
WB and ELISA. Suggested dilution of 1:1,000 to 1:4,000. To minimise background staining, a higher concentration of detergent (such as Tween-20) is suggested in the dilution and washing steps. Biosensis recommends that the optimal working dilution should be determined by the end user.

Type: Primary
Antigen: PS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: [delete]
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR