Human Recombinant Myostatin (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
Myostatin, also known as GDF-8, is a conserved member of the TGF-beta superfamily. Myostatin is an essential regulator of skeletal muscle mass and cardiac muscle development and function. Myostatin is a secreted protein that negatively regulates skeletal muscle growth by determining muscle fiber number and size.
- High quality
- Low endotoxin
- Affordable
Myostatin binds one of the two activin type II receptors (ACTRIIA or ACTRIIB) to activate SMAD signaling. Myostatin also activates MAPK signaling through TAK1-MKK6 and Ras pathways. Inhibition of myostatin increases muscle mass in a number of human disease animal models, such as muscular dystrophy.
Caution: For research use only.