Human Recombinant RANK LIGAND (from E. coli)

Supplier: FUJIFILM IRVINE SCIENTIFIC, INC

100-19-100UG 100-19-10UG 100-19-1MG 100-19-500UG
10788-990EA 1368.83 CAD
10788-990 10788-992 10788-994 10788-996 10821-890
Human Recombinant RANK LIGAND (from E. coli)
Proteins and Peptides

Interleukin-1 receptor antagonist (IL-1RA) binds the interleukin 1 receptor (IL-1R) to antagonize interleukin 1 alpha and beta signaling. IL-1RA is secreted by immune cells, epithelial cells, hepatocytes, and adipocytes to inhibit IL-1-induced inflammation. IL-1RA therapy is used to treat auto-inflammatory diseases such as diabetes and rheumatoid arthritis.


  • High quality
  • Low endotoxin
  • Affordable


Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.


Caution: For research use only.

Order Now

SPECIFICATIONS

Protein/peptide name RANK LIGAND
Environmentally Preferable
Protein synonyms OPGL|TNFSF11|ODF|TRANCE
Protein/peptide type Recombinant
Protein Function Cytokine
Species Human
Source E. coli
Conjugation Unconjugated
Gene ID O14788
Sequence EKAMVDGSWL DLAKRSKLEA QPFAHLTINA TDIPSGSHKV SLSSWYHDRG WAKISNMTFS NGKLIVNQDG FYYLYANICF RHHETSGDLA TEYLQLMVYV TKTSIKIPSS HTLMKGGSTK YWSGNSEFHF YSINVGGFFK LRSGEEISIE VSNPSLLDPD QDATYFGAFK VRDID
Protease-free N
Animal-free N
Nuclease-free N
Carrier-free Y
Endotoxin-free N
Endotoxin level Low
Endotoxin content Endotoxin LAL (EU/µg) ≤1
UniProtKB O14788
Biological activity Bioactivity assay: Ability to induce bioluminescence in engineered RANKL bioassay cells , ED50 ≤50 ng/ml, bioactivity ≥2.0×10⁴ units/mg
Purity ≥95%
Grade RUO
Formulation 10 mM sodium phosphate, pH 7.5
Molecular weight 19.7 kDa
Reconstitution instructions Sterile water at 0.1 mg/ml
Storage conditions −20 °C
Shipping temperature RT
Tested applications Bioassay

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR