Human Recombinant Fractalkine (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
100-174-100UG 100-174-1MG 100-174-20UG 100-174-5UG 100-174-500UG
10788-958EA 884.72 CAD
10788-958 10788-960 10788-962 10788-964 10821-986
Human Recombinant Fractalkine (from E. coli)
Proteins and Peptides
Fractalkine, also known as CX3CL1, is a cytokine protein containing a CX3C chemokine motif atop a mucin stalk. Fractalkine is produced by non-hemopoietic cells, including neurons and astrocytes. Soluble fractalkine functions as a chemoattractant for T cells and monocytes. Cell-membrane-bound fractalkine, which is induced on activated endothelial cells, promotes leukocyte adhesion. The transmembrane chemokine receptor CX3CR1 mediates the adhesive and chemoattractant functions of fractalkine.
  • High quality
  • Low endotoxin
  • Affordable
Caution: For research use only.