Human Recombinant GROgamma/ MIP-2beta/ CXCL3 (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
Growth regulated protein gamma (GRO-gamma), also called CXCL3, acts through the chemokine receptor CXCR2 to promote monocyte migration and adhesion. GRO-gamma also induces the migration of cerebellar granule neuron precursor cells.
Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.
A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions. This product is for research use only and is not for use in humans.
Ordering information: Ships at room temperature; Store at 20°C upon delivery.