Human Recombinant IL-29 (from E. coli)

Supplier: FUJIFILM IRVINE SCIENTIFIC, INC

100-169-100UG 100-169-1MG 100-169-20UG 100-169-5UG 100-169-500UG
10788-902EA 884.72 CAD
10788-902 10788-904 10788-906 10788-908 10822-218
Human Recombinant IL-29 (from E. coli)
Proteins and Peptides

Interleukin 29 (IL-29), also known as IFN-λ, is a type III interferon produced by virally infected cells. IL-29 plays an important role in host defenses against microbes and antiviral activity. IL-29 shares homology with interleukin 28 (IL-28) and binds the class II cytokine receptor IL-28R.


  • High quality
  • Low endotoxin
  • Affordable


Caution: For research use only.

Order Now

SPECIFICATIONS

Protein/peptide name IL-29
Environmentally Preferable
Protein synonyms IFN-lambda|Interferon lambda|Type III interferon|IFNλ1
Protein/peptide type Recombinant
Protein Function Cytokine
Species Human
Source E. coli
Conjugation Unconjugated
Gene ID Q8IU54
Sequence MPTSKPTTTG KGCHIGRFKS LSPQELASFK KARDALEESL KLKNWSCSSP VFPGNWDLRL LQVRERPVAL EAELALTLKV LEAAAGPALE DVLDQPLHTL HHILSQLQAC IQPQPTAGPR PRGRLHHWLH RLQEAPKKES AGCLEASVTF NLFRLLTRDL KYVADGNLCL RTSTHPEST
Protease-free N
Animal-free N
Nuclease-free N
Carrier-free Y
Endotoxin-free N
Endotoxin level Low
Endotoxin content Endotoxin LAL (EU/µg) ≤1
Biological activity Bioactivity: No biological activity data is available at this time
Purity ≥95%
Grade RUO
Formulation 0.1% Trifluoroacetic acid (TFA)
Molecular weight 19.9 kDa
Reconstitution instructions Sterile water at 0.1 mg/ml
Storage conditions –20 °C
Shipping temperature RT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR