Human Recombinant IL-1B (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
Interleukin 1 beta (IL-1β) is a pro-inflammatory cytokine that is produced by monocytes, macrophages, and dendritic cells (DCs). IL-1β signals through the IL-1 receptor, type 1 (IL-1R1) to activate the myeloid differentiation primary response 88 (MYD88) signaling pathway, which contains the cytoplasmic Toll/IL-1 receptor (TIR) domain adapter.
- High quality
- Low endotoxin
- Affordable
IL-1β and the independently regulated IL-1α protein have overlapping pro-inflammatory activities to induce adhesion molecule expression on epithelial cells, control fever induction, initiate rheumatoid arthritis, and promote septic shock.
Caution: For research use only.