Human Recombinant TSLP (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
100-165-100UG 100-165-10UG 100-165-1MG 100-165-500UG
10787-434EA 1368.83 CAD
10787-434 10788-716 10788-718 10788-720 10822-414
Human Recombinant TSLP (from E. coli)
Proteins and Peptides
Thymic stromal lymphopoietin (TSLP) is a hematopoietic cytokine produced in several tissues including the heart, liver, and prostate. TSLP induces the release of T-cell attracting chemokines from monocytes, and regulates the maturation of myeloid and epidermal dendritic cells. TSLP signals through a heterodimeric receptor complex containing the TSLP receptor (TSLP-R/CRLF2) and IL-7R alpha chain.
  • High quality
  • Low endotoxin
  • Affordable
Caution: For research use only.