Mouse Recombinant IL-7 (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
200-03-10UG 200-03-1MG 200-03-100UG 200-03-500UG
10787-844EA 354.2 CAD
10787-844 10787-846 10787-848 10789-078 10822-124
Mouse Recombinant IL-7 (from E. coli)
Proteins and Peptides
Interleukin 7 (IL-7) is a hematopoietic cytokine that is an important regulator of B and T cell development. IL-7 is secreted by bone marrow and thymic stromal cells, dendritic cells, intestinal epithelial cells, hepatocytes, and keratinocytes. IL-7 signals through the interleukin 7 receptor (IL-7R) to promote the differentiation of hematopoietic stem cells into T cells, B cells, and natural killer cells. IL-7 is also a regulator of intestinal mucosal lymphocyte proliferation. Human and mouse IL-7 show species cross-reactivity.
  • High Quality
  • Low Endotoxin
  • Affordable
A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.  This product is for research use only and is not for use in humans.  
Caution: For research use only.