Human Recombinant Galectin-3 (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
Galectin-3 belongs to the lectin family of carbohydrate binding proteins. Galectin-3 is expressed by a wide range of cell types including activated T cells, tumor cells, macrophages, osteoclasts, fibroblasts, and epithelial cells. Galectin-3 has specific binding affinity for beta-galactoside sugar moieties and has functional roles during development, innate immunity, cell apoptosis, and tumor metastasis. Galectin-3 is associated with cancer, heart failure, stroke, and inflammation. The amino acid sequences of human and mouse Galectin-3 proteins share 80% homology.
Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.
A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions. This product is for research use only and is not for use in humans.
Ordering information: Ships at room temperature; Store at 20°C upon delivery.