Human Recombinant IL-21 (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
100-90-100UG 100-90-10UG 100-90-1MG 100-90-500UG
10787-802EA 1368.83 CAD
10787-802 10787-804 10787-806 10787-808 10822-196
Human Recombinant IL-21 (from E. coli)
Proteins and Peptides
Interleukin 21 (IL-21) is a member of the common-gamma chain family of cytokines that are involved in immunoregulation. IL-21 is normally expressed by activated CD4+ T cells and is aberrantly expressed in Hodgkin lymphoma cells. The IL-21 receptor (IL-21R) activates the JAK/STAT signaling pathway and is expressed on T, B, and natural killer (NK) cells.
  • High quality
  • Low endotoxin
  • Affordable
Within the B cell lineage, IL-21 is a switch factor regulating IgG1 and IgG3 antibody production. IL-21 also cooperates with IL-4 for the production of multiple antibody classes in B cells. IL-21 has pleiotropic effects on the proliferation, differentiation, and effector functions of B, T, NK, and dendritic cells.
Caution: For research use only.