Human Recombinant IL-20 (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
100-89-100UG
100-89-10UG
100-89-1MG
100-89-500UG
10787-794EA
1256.31
CAD
10787-794
10787-796
10787-798
10787-800
10822-194
Human Recombinant IL-20 (from E. coli)
Proteins and Peptides
Interleukin 20 (IL-20) is structurally related to interleukin 10 (IL-10) and is produced by keratinocytes and monocytes. IL-20 acts through the STAT3 signaling pathway to regulate the proliferation of keratinocytes during epidermal inflammation.
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.