Human Recombinant FGF-Basic 154 (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
100-146-50UG 100-146-10UG 100-146-1MG 100-146-500UG 100-146-100UG
10787-332EA 358.96 CAD
10787-332 10788-696 10788-698 10821-764 10821-972
Human Recombinant FGF-Basic 154 (from E. coli)
Proteins and Peptides
Basic fibroblast growth factor (FGF-basic), also known as FGF-2, is expressed by endothelial cells and is a mediator of angiogenesis. FGF-basic also has cardioprotective functions during heart injury. FGF-basic is a critical component for embryonic stem cell culture systems and is necessary for maintaining cells in an undifferentiated state. Recombinant FGF-basic 154 is the full length FGF-basic protein encoded by the human FGF-2 gene. FGF-basic 154 is the most popular tissue culture product at Shenandoah Biotechnology, Inc. There are no detectable differences in biological activity between FGF-basic 154 and the truncated FGF-basic 147 proteins.
  • High quality
  • Low endotoxin
  • Affordable
Caution: For research use only.