Human Recombinant Myostatin Propeptide (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
100-142-100UG
100-142-1MG
100-142-25UG
100-142-5UG
100-142-500UG
10787-312EA
1368.83
CAD
10787-312
10787-314
10787-316
10787-318
10822-306
Human Recombinant Myostatin Propeptide (from E. coli)
Proteins and Peptides
Myostatin (GDF-8), a member of the TGF-beta superfamily, is a potent and negative regulator of skeletal muscle mass. The recombinant myostatin propeptide binds and inhibits myostatin to enhance the repair and regeneration of injured muscle and bone.
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.