Human Recombinant MCP-3 (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
100-131-100UG 100-131-10UG 100-131-1MG 100-131-500UG
10788-854EA 884.72 CAD
10788-854 10788-856 10788-858 10788-860 10822-256
Human Recombinant MCP-3 (from E. coli)
Proteins and Peptides
Monocyte chemotactic protein 3 (MCP-3), also called CCL7, is produced by macrophages and tumor cell lines. MCP-3 signals through the G protein-coupled receptors CCR1, CCR2, and CCR3. MCP-3 chemoattracts monocytes and regulates macrophage function during inflammation and metastasis.
  • High quality
  • Low endotoxin
  • Affordable
Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.
Caution: For research use only.