Human Recombinant IL-2 (from E. coli)

Supplier: FUJIFILM IRVINE SCIENTIFIC, INC

100-12-10UG 100-12-1MG 100-12-50UG 100-12-500UG 100-12-100UG
10787-214EA 143.42 CAD
10787-214 10787-216 10787-218 10821-834 10822-082
Human Recombinant IL-2 (from E. coli)
Proteins and Peptides

Interleukin 2 (IL-2) is an immunomodulatory cytokine that is produced by lymphocytes.  IL-2 signals through the IL-2R receptor to induce activated T cell proliferation and promote T cell differentiation. IL-2 also stimulates the proliferation and differentiation of B cells, natural killer cells, monocytes, and macrophages.


  • High quality
  • Low endotoxin
  • Affordable


Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.


Caution: For research use only.

Order Now

SPECIFICATIONS

Protein/peptide name IL-2
Environmentally Preferable
Protein synonyms TCGF|T cell growth factor|aldesleukin
Protein/peptide type Recombinant
Protein Function Cytokine
Species Human
Source E. coli
Conjugation Unconjugated
Gene ID P60568
Sequence MAPTSSSTKK TQLQLEHLLL DLQMILNGIN NYKNPKLTRM LTFKFYMPKK ATELKHLQCL EEELKPLEEV LNLAQSKNFH LRPRDLISNI NVIVLELKGS ETTFMCEYAD ETATIVEFLN RWITFCQSII STLT
Protease-free N
Animal-free N
Nuclease-free N
Carrier-free Y
Endotoxin-free N
Endotoxin level Low
Endotoxin content Endotoxin LAL (EU/µg) ≤0.1
UniProtKB P60568
Biological activity Bioactivity assay: CTLL-2 cell proliferation, ED50 ≤3 ng/ml, bioactivity ≥3.3×10⁵ units/mg
Purity ≥97%
Grade RUO
Formulation 0.1% Trifluoroacetic Acid (TFA)
Molecular weight 15.5 kDa
Reconstitution instructions Sterile 10 mM HCl at 0.1 mg/ml
Storage conditions −20 °C
Shipping temperature RT
Tested applications Bioassay

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR