CritiCore Taffeta-Hypalon/CSM Shoe Covers
Supplier: CritiCore Protective Wear
CritiCore
10798-894 10798-886 10798-888 10798-890 10798-958 10798-892
10798-958PK 42.65 CAD
10798-958 10798-888 10798-886 10798-894 10798-892 10798-890
CritiCore Taffeta-Hypalon/CSM Shoe Covers
Workwear
These reusable cleanroom shoe covers are manufactured with a soft, tan CSM (Hypalon) sole and 100% monofilament polyester taffeta uppers.
  • Tunneled elastic at the ankle
  • Color-coded loop for size identification
  • Single snap adjustment on the back
Extremely lightweight shoe covers are available in white only.
Ordering information: Contact your Avantor sales representative for more information.
Packaging: Shoe covers are packaged in pairs.