Aluminium Weighing Dishes, Dyn-A-Med
Supplier: Dyn-A-Med
80064 80065 80065L
11300-445CS 346.19 CAD
11300-445 11300-450 11300-455
Aluminium Weighing Dishes, Dyn-A-Med
Boats Weighing Boats
Disposable aluminum foil dishes used for weighing or moisture testing.
  • Used in wastewater, chemical, pharmaceutical, educational and dairy applications
Available with crimped sides, flat smooth bottom and a finger tab handle on the rim or totally smooth with no finger tab.