VWR® Volumetric Pipettes, Reusable, Color Coded, Class A
Supplier: VWR International
10546-004 10546-006 10546-008 10546-010 10546-012 10546-014 10546-016 10546-018 10546-020 10546-022 10546-024 10546-026 10546-028 10546-030 10546-032 10546-034 10546-036 10546-038
10546-004CS 290.81 CAD
10546-004 10546-006 10546-008 10546-010 10546-012 10546-014 10546-016 10546-018 10546-020 10546-022 10546-024 10546-026 10546-028 10546-030 10546-032 10546-034 10546-036 10546-038
VWR® Volumetric Pipettes, Reusable, Color Coded, Class A
Pipettes Serological Pipettes
Made from ASTM E438 Type 1 Class A borosilicate 33 expansion glass and as per ASTM specification E969.
  • Reusable
  • Color-coded for easy sorting
  • Borosilicate glass fabrication
Calibrated at 20 °C TD-EX. These pipettes are unserialized or batch certified.
Certifications: Pipettes are color coded as per ASTM specification E1273.