VWR® Volumetric Flask with PE Stopper, Class A
Supplier: VWR International
10545-984 10545-986 10545-988 10545-990 10545-992 10545-994 10545-996 10545-998 10546-000 10546-002
10545-984CS 582.44 CAD
10545-984 10545-986 10545-988 10545-990 10545-992 10545-994 10545-996 10545-998 10546-000 10546-002
VWR® Volumetric Flask with PE Stopper, Class A
Flasks Volumetric Flasks
These volumetric flasks are batch certified and are calibrated at 20 degrees Celsius To Contain.
  • Borosilicate glass fabrication
  • Polyethylene stopper construction
  • Durable white marking spot
They feature a single graduation ring and polyethylene stopper with marking spot.
Made from ASTM E438 Type 1 Class A Borosilicate 33 expansion glass as per ASTM specification E237 and E288. Durable white color printing which has good life in laboratory environment.