Human Recombinant TNF-A (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
100-111-10UG 100-111-1MG 100-111-50UG 100-111-500UG 100-111-100UG
10787-180EA 141.54 CAD
10787-180 10787-182 10787-184 10821-910 10822-396
Human Recombinant TNF-A (from E. coli)
Proteins and Peptides
Tumor necrosis factor alpha (TNF-α) is an inflammatory cytokine secreted by macrophages, monocytes, neutrophils, T cells, and NK-cells following stimulation by bacterial lipopolysaccharide (LPS).
  • High quality
  • Low endotoxin
  • Affordable
TNF-α signal activation occurs through two receptors, TNFR1 and TNFR2. TNFR1 is expressed on most cell types, unlike TNFR2, which is expressed mainly on immune cells. TNF-α functions to stimulate phagocytosis in macrophages, chemoattract neutrophils, increase insulin resistance, and induce fever.
Caution: For research use only.