Human Recombinant IL-7 (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
100-11-100UG 100-11-10UG 100-11-1MG 100-11-500UG
10787-172EA 1256.7 CAD
10787-172 10787-174 10787-176 10787-178 10822-122
Human Recombinant IL-7 (from E. coli)
Proteins and Peptides
Interleukin 7 (IL-7) is a hematopoietic cytokine that is an important regulator of B and T cell development. IL-7 is secreted by bone marrow and thymic stromal cells, dendritic cells, intestinal epithelial cells, hepatocytes, and keratinocytes. IL-7 signals through the interleukin 7 receptor (IL-7R) to promote the differentiation of hematopoietic stem cells into T cells, B cells, and natural killer cells. IL-7 is also a regulator of intestinal mucosal lymphocyte proliferation. Human and mouse IL-7 show species cross-reactivity.
  • High quality
  • Low endotoxin
  • Affordable
Caution: For research use only.