Properties Of Matter: What Is Oobleck?
Supplier: Ward's Science
IS1003
470163-310KT 63.85 CAD
470163-310
Properties Of Matter: What Is Oobleck?
Educational Materials Physics Educational Materials Properties of Matter Learning Activities in Physics
Hands On Science For Elementary Students!
  • Ideal for grades K-4
  • Introductory hands on activity
  • Non-hazardous chemicals included
This kit has been designed to introduce young students (K-4) to the concepts of matter in a fun, safe, hands-on way. Students are given samples of both a solid and a liquid so that they can perform some simple experiments that demonstrate the differences. The students are then given a sample of “Our Friend Oobleck” (a non-Newtonian fluid) to use in the same experiments and are asked to determine if Oobleck is a solid or a liquid. 
Ordering information: Includes Oobleck Powder, Solid Samples, Holding Trays, Testing Spatulas, and Teacher's Guide and Student Copymaster.