Human Recombinant Resistin (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
100-107-100UG 100-107-1MG 100-107-25UG 100-107-5UG 100-107-500UG
10787-148EA 1256.7 CAD
10787-148 10787-150 10787-152 10787-154 10822-352
Human Recombinant Resistin (from E. coli)
Proteins and Peptides
Resistin is a peptide hormone belonging to a class of cysteine-rich secreted proteins, termed the resistin-like molecules (RELM) family. Resistin is produced by macrophages and functions during insulin sensitivity and inflammatory processes.
  • High Quality
  • Low Endotoxin
  • Affordable
A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.  This product is for research use only and is not for use in humans.  
Caution: For research use only.