VWR® Test Tubes with Sturdy Lip, Borosilicate Glass
Supplier: VWR
097.415.06 097.415.07 097.415.03 097.415.01 097.415.02 097.415.08 097.415.04 097.415.08A 097.415.05
10545-916CS 943.95 CAD
10545-916 10545-928 10545-918 10545-930 10545-920 10545-922 10545-924 10545-914 10545-926
VWR® Test Tubes with Sturdy Lip, Borosilicate Glass
Tubes Test Tubes
High quality test tubes designed and constructed for optimal performance.
  • High quality borosilicate glass construction
  • Durable white ceramic enamel marking spot
Allows use at temprature up to 500 °C and have excellent resistance to chemcial attack.
Tubes feature a sturdy lip and uniform wall thickness for maximum heat transfer and chemical resistance.
Certifications: Made from ASTM 438 Type 1 Class A borosilicate 33 expansion glass and as per ASTM specification E982.