Human Recombinant M-CSF (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
100-03-100UG 100-03-10UG 100-03-1MG 100-03-500UG
10788-832EA 1051.58 CAD
10788-832 10788-834 10788-836 10788-838 10822-264
Human Recombinant M-CSF (from E. coli)
Proteins and Peptides
Macrophage colony stimulating factor (M-CSF) is a hematopoietic growth factor that is widely produced by a variety of cells. M-CSF stimulates the proliferation and differentiation of hematopoietic stem cells into monocyte and macrophage cell types.
  • High quality
  • Low endotoxin
  • Affordable
M-CSF also acts through the colony stimulating factor 1 receptor (CSF1R) to modulate processes involved in immunology, bone metabolism, fertility, and pregnancy. Human M-CSF shows activity on mouse cells; however, mouse M-CSF shows no activity on human cells.
Caution: For research use only.