SP Wilmad-LabGlass Plastic Clamps, SP Industries
Supplier: SP Industries
LG-1004-106
14200-038EA 9.09 CAD
14200-038
SP Wilmad-LabGlass Plastic Clamps, SP Industries
Clamps Standard Taper and Spherical Joint Clamps
Color coded by size, these clamps hold Standard Taper joints in place securely under light pressure.
  • Made of the plastic resin polyoxymethylene (POM), they can withstand temperatures up to 150°C
  • Resistant to concentrated alkalais and dilute acids, but not resistant to concentrated acids or halogen gases