Antistatic Polystyrene Pour Boats, Simport Scientific
Supplier: Simport Scientific
D251-2 D251-3 D251-1
CA11016-310CS 70.34 CAD
CA11016-310 CA11027-122 76299-240
Antistatic Polystyrene Pour Boats, Simport Scientific
Boats Weighing Boats
These three sizes of antistatic polystyrene pour boats are specially made to facilitate dispensing of powdered and liquid materials.
  • Smooth, uniform and economical
The molded material used is thicker than conventional weighing dishes. They can be used safely to weight static-affected materials, and will withstand temperatures up to 80°C.