Hunter Fire Assay Cupels, MGO, Anachemia
Supplier: Anachemia
ANACCUPM8 ANACCUPM9 ANACCUPM5-D ANACCUPM5A-D
10119-364CS 170.56 CAD
10119-364 10119-356 10119-334 10119-346
Hunter Fire Assay Cupels, MGO, Anachemia
Crucibles
Hunter cupels have been a consistent mainstay in the assay industry for over 30 years
  • Consistent industry mainstay
  • For use in precious metal refinement
  • Variety of sizes available
Used by precious metals refining and assay establishments worldwide, Hunter cupels are manufactured to exact and consistent standards using high purity magnesium oxide. They are available in a variety of different sizes.