VWR® Round-Bottom Boiling Flasks
Supplier: VWR International
10536-652 10536-654 10536-664 10536-666 10536-668 10536-670 10536-672 10536-674
10536-652CS 315.34 CAD
10536-652 10536-654 10536-664 10536-666 10536-668 10536-670 10536-672 10536-674
VWR® Round-Bottom Boiling Flasks
Flasks Round Bottom Flasks
Durable boiling flasks have good temperature and chemical resistance.
  • Good temperature resistance
  • Chemically resistant
  • Uniform wall thickness
Manufactured from ASTM E438 Type 1 Class A borosilicate 33 expansion glass. As per ASTM specification E 1403 Type 1. The flasks feature uniform wall thickness, which makes them ideal for heating.