Freezing Point Depression
Supplier: ALDON CORP SE
IS8014
470163-094KT 81.2 CAD
470163-094
Freezing Point Depression
Educational Materials Chemistry Educational Materials Properties of Matter Learning Activities in Chemistry
How do different solvents affect the freezing point?
  • Students determine freezing point depression
  • Classic AP chemistry topic
Molecular mass is a parameter, which is useful in determining the identity of an unknown compound. One technique to determine the molecular mass of an unknown is to measure the effect the compound has on the freezing point of a solvent in which the unknown is dissolved. The freezing point of a solution is a colligative property. That is, it is a property which varies based on the number of particles (solute) dissolved in the solvent and not on the chemical makeup of the particles themselves. Other colligative properties, which also can be used to determine molecular mass, are osmotic pressure, vapor pressure and boiling point. The nonpolar solvent 2,6-di-tert-butyl-4-methylphenol has a freezing point of approximately 70°C. A quantity of para-dichlorobenzene will be dissolved in the solvent and the effect on the freezing point determined. The freezing point depression constant will be calculated for the solvent. The experiment will be repeated with each of two unknowns and the molecular weight of the unknowns will be determined from the freezing point depression.
Ordering information: Lab includes Di-Tert-Butyl-4-Methylphenol Crystals, p-Dichlorobenzene Crystals, Stearic Acid Flakes, and Naphthalene Flakes.