Corning® Preassembled Closed System Solutions for Erlenmeyer Flasks, 5 L, Corning
Supplier: Corning
11500 11501 11502
10790-808CS 688.5 CAD
10790-808 10790-810 10790-812
Corning® Preassembled Closed System Solutions for Erlenmeyer Flasks, 5 L, Corning
Flasks Erlenmeyer Flasks
Closed cell culture systems help reduce the risk of adventitious contamination during drug development and manufacturing.
  • Double bagged for GMP applications
  • Animal component free vessels
  • Wide range of product offerings
  • Customizable solutions
The internal end is a rigid polypropylene dip tube designed to reach the bottom of the flask. The venting port ends in a 50 mm diameter vent filter with a 0.2 μm pore PVDF membrane and a barbed fitting. The liquid transfer port uses either a 1/4" ID (3/8" OD) chemically resistant, heat sealable flexible tubing on the external side of the cap ending in a male plastic coupler (MPC) male disconnect with a sealing cap or a 1/8" ID (1/4" OD) chemically resistant, heat sealable flexible tubing on the external side of the cap ending in a male Luer lock (MLL) closed with a female Luer cap.
Ordering components and assembling tubing sets in-house can add complexity and time to your processes.
Tube length: 36"
Certifications: BSE/TSE compliant accessories. SAL 10-6 sterilization.
Caution: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.