Quantofix QUAT Test Strips, CTL Scientific Supply
Supplier: CTL Scientific Supply
91337
10217-572EA 92.97 CAD
10217-572
Quantofix QUAT Test Strips, CTL Scientific Supply
Environmental Test Kits
Quantofix QUAT test strips for the rapid and reliable determination of quaternary ammonium compounds (QUAT) in solutions.
Quantofix Quat has a gradation of scale: 0-10-25-50-100-250-500-1000 mg/L.
Ordering information: Packaged as a box of 100.