Quantofix Sulfate Test Strips, CTL Scientific Supply
Supplier: CTL Scientific Supply
91329
10217-558EA 101.01 CAD
10217-558
Quantofix Sulfate Test Strips, CTL Scientific Supply
Environmental Test Kits
Semi-quantitative test strips for the determination of Sulfate. Ideal for rapid and easy analysis of sulfate directly on-site.
  • No maintenance or calibration is required.
Ordering information: Packaged as a box of 100.