Bucket Liners, Micronova
Supplier: Micronova
BKL7-3119IR BKL7-3119 BKL1-1818IR BKL-2424IR BKL-2430IR BKL993521IRB
68400-780CS 468.45 CAD
68400-780 89173-176 89187-734 89426-906 89426-908 75873-828
Bucket Liners, Micronova
Bags Waste Disposal Bags
The unique LDPE/Nylon laminate is a superb barrier material for lining totes to transport raw materials or to line buckets containing disinfectants or other cleaning solutions.
  • Laminate is 75% LDPE, 25% nylon
  • Excellent for disinfectants and cleaning solutions
  • Ideal for product parts or raw material transfer
  • Thickness of 3 mil
  • Will hold up to autoclaving with liquid contents
  • Available sterile (irradiated) or non sterile
Non sterile liners are bulk packed. Select sterile liners are available bulk packed.
Packaging: Flat packed, either individually double-bagged or in bulk packs. Packed in a case of 24 or case of 72 for bulk pack.