Quantofix Peroxide Test Strips, CTL Scientific Supply
Supplier: CTL Scientific Supply
91333 91312 91319
10217-538EA 92.26 CAD
10217-538 10217-536 10217-540
Quantofix Peroxide Test Strips, CTL Scientific Supply
Environmental Test Kits
Quantofix Peroxide for the semi-quantitative determination of hydrogen peroxide (H2O2) and peroxides in solutions.
Color changes with H2O2 from white to blue.
Ordering information: Packaged as a box of 100.