Quantofix Nitrate/Nitrite Test Strips, CTL Scientific Supply
Supplier: CTL SCIENTIFIC SUPPLY CORP
Quantofix Nitrate/Nitrite test strips are for the rapid and reliable determination of Nitrate and Nitrite in solutions
Quantofix Nitrate 100; This test is the highly sensitive version of Quantofix Nitrate/Nitrite. This test allows to detect nitrate down to 5 mg/L. The warning pad for nitrite detects concentrations down to 0.5 mg/L. This test is especially useful for the dairy industry where low levels of nitrate need to be monitored.
Color change is white to red-violet. Quantofix Nitrate 100 has a gradation of scale: 0-5-10-25-50-75-100 mg/L.
The easy dip and read procedure provides a reliable result within 1 minute.
Ordering information: Packaged as a box of 100.