Quantofix Copper Test Strips, CTL Scientific Supply
Supplier: CTL Scientific Supply
91304
10217-512EA 84.66 CAD
10217-512
Quantofix Copper Test Strips, CTL Scientific Supply
Environmental Test Kits
Quantofix Copper Test strips for the semi-quantitative determination of copper (Cu + /Cu 2+) in solutions.
Quantofix Copper Test strips has a gradation of scale: 0-10-30-100-300 mg/L.
Ordering information: Packaged as a box of 100.