QUS Series Ultra Stack and Hang Bins, Quantum Storage Systems
Supplier: Quantum Storage Systems
QUS210BL QUS250IV QUS240BL QUS240RD QUS250YL QUS220RD QUS210RD QUS230RD QUS220BL QUS250BL QUS230YL QUS220YL QUS250GN QUS210YL QUS240YL QUS250RD QUS250BK QUS230BL
76202-856CS 315.54 CAD
76202-856 76202-858 10752-458 10752-468 10752-456 10752-478 10752-466 76202-860 10752-454 10752-476 10752-464 10752-452 10752-474 10752-462 10752-472 10752-460 10752-470 10752-480
QUS Series Ultra Stack and Hang Bins, Quantum Storage Systems
Boxes Dispenser Boxes
The QUS200 Series Ultra stack and hang bin is made from strong and durable injection molded plastic.
  • Available in blue, yellow, red
  • Autoclavable up to 250°F
  • Resistant to extreme cold
  • Large molded front label slot for ID label
Built-in rear hanger allows bins to hang from louvered panels of rails. Dividers maximize flexiblity and keep contents organized.