Quantofix Arsenic Test Kits, CTL Scientific Supply
Supplier: CTL SCIENTIFIC SUPPLY CORP
91334 91332 91345
10217-502EA 259.07 CAD
10217-502 10217-504 10217-506
Quantofix Arsenic Test Kits, CTL Scientific Supply
Environmental Test Kits
Quantofix Arsenic test kits and reagents for the semi-quantitative determination of arsenic in solutions
Quantofix Arsenic 10 has a gradation of scale: 0-0.01-0.025-0.05-0.1-0.5 ppm.
The addition of zinc and a solid acid results in the formation of arsine from any arsenic compounds in the sample solution. Arsine reacts in the gaeous phase with the mercury bromide in the reaction zone of the test stick to form mixed arsenic/mercury halides, which, depending on the concentration, are colored yellow to brown.
Ordering information: Packaged as a box of 100, strips and reagent.
Order Now
SPECIFICATIONS
Environmentally Preferable
Storage RT