Quantofix Carbonate Hardness Test Strips, CTL Scientific Supply
Supplier: CTL Scientific Supply
91323
10217-496EA 96.44 CAD
10217-496
Quantofix Carbonate Hardness Test Strips, CTL Scientific Supply
Environmental Test Kits
Quantofix Carbonate Hardness Test strips for the semi-quantitative determination of the carbonate hardness
Quantofix Carbonate Hardness Test strips have a gradation of scale: 0-3-6-10-15-20 mg/L light green to blue. Strong bases or other acid-consuming substances cause false results.
Carbonate hardness is that part of the total hardness resulting from the content of alkaline earth ions, which are equivalent to the carbonate and hydrogen carbonate ions present in the water. Thus the carbonate hardness is the portion of the total hardness which contributes to the formation of alkaline earth carbonates (boiler scale).
Ordering information: Packaged as a box of 100.