Veggie-Bites™ Guinea Pig Enrichment Treats, Certified

Supplier: Bio-Serv
F5158 F5158 S5158-1 F5158-1 F5158-1
CA89067-444EA 526.87 CAD
CA89067-444 89067-444 76406-494 76419-518 CA76419-518
Veggie-Bites™ Guinea Pig Enrichment Treats, Certified
Animal Supplies Animal Feed
An all-natural, crunchy vegetable treat that is a medley of dehydrated sliced carrots, puffed peas and whole kernel corn.

  • Less labor intensive and more sanitary than perishable vegetables
  • Feed as a training reward or foraging treat
  • Nutritionally assayed
  • Shelf life: 12 months, cool dry storage

Ingredients: Dehydrated carrots, puffed peas, dried whole kernel corn.

Caloric value: 3.44 kcal/gm
Nutritional ash content: 2.0%
Nutritional carbohydrate content: 67.0%
Nutritional fat content: 4.0%
Nutritional fibre content: 5.0%
Nutritional moisture content: <12%
Nutritional protein content: 10.0%

Certifications: Certified (Contaminant screened).

Ordering information: Product is no longer gamma irradiated. It is now Certified. If gamma irradiation is required, please contact customer service for pricing and lead time.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR